Skip to Content
Merck
All Photos(1)

Key Documents

W0125

Sigma-Aldrich

Wheat germ Untreated

suitable for enzyme extraction

Sign Into View Organizational & Contract Pricing

Select a Size

5 KG
€146.00

€146.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 KG
€146.00

About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

€146.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

wheat

Quality Level

form

flakes

suitability

suitable for enzyme extraction

storage temp.

2-8°C

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Englert et al.
Nucleic acids research, 33(1), 388-399 (2005-01-18)
Pre-tRNA splicing is an essential process in all eukaryotes. It requires the concerted action of an endonuclease to remove the intron and a ligase for joining the resulting tRNA halves as studied best in the yeast Saccharomyces cerevisiae. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service