Skip to Content
Merck
All Photos(4)

Documents

S7796

Sigma-Aldrich

Sisomicin sulfate salt

≥580 μg/mg (USP)

Synonym(s):

O-3-Deoxy-4-C-methyl-3-(methylamino)-beta-L-arabinopyranosyl-(1->4)-O-(2,6-diamino-2,3,4,6-tetradeoxy-alpha-D-glycero-hex-4-enopyranosyl-(1->6))-2-deoxy-L-streptamine sulfate (2:5) (salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H37N5O7 · 2.5H2O4S
CAS Number:
Molecular Weight:
692.72
EC Number:
UNSPSC Code:
51281665
PubChem Substance ID:
NACRES:
NA.85

form

powder

concentration

≥580 μg/mg (USP)

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@H](OC[C@]1(C)O)O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3OC(CN)=CC[C@H]3N)[C@@H]2O.CN[C@@H]4[C@@H](O)[C@H](OC[C@]4(C)O)O[C@H]5[C@H](N)C[C@H](N)[C@@H](O[C@H]6OC(CN)=CC[C@H]6N)[C@@H]5O

InChI

1S/2C19H37N5O7.5H2O4S/c2*1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17;5*1-5(2,3)4/h2*3,9-18,24-27H,4-7,20-23H2,1-2H3;5*(H2,1,2,3,4)/t2*9-,10+,11-,12+,13-,14-,15+,16-,17-,18-,19+;;;;;/m11...../s1

InChI key

CIKNYWFPGZCHDL-ZHFUJENKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Sisomicin is an aminoglycoside antibiotic isolated from Micromonopora inyoensis. It is effective against most strains of Klebsiellk spp., Escherichia coli, P. aeruginosa, Enterobacter and Proteus spp.. It is a potential treatment for conjunctiva . It is used in disk and tube dilution sensitivity test.

Biochem/physiol Actions

Sisomicin interferes with protein synthesis at the level of functional ribosome assembly. The mechanism is similar to that of gentamycin. Sisomicin closely resembles gentamicin but is more effective against Pseudomonas aeruginosa and indole-positive Proteus.

Packaging

250MG,1G,5G

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Waitz
Antimicrobial agents and chemotherapy, 4(4), 445-454 (1973-10-01)
Poor statistical correlation was obtained between tube dilution test results and disk test results by using standard procedures. Significant regressions were obtained although they were not linear. Different regressions were obtained with different bacterial species. It is suggested that for
Sławomir Wilczyński et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 45(3), 251-262 (2011-11-26)
Radiation sterilization technology is more actively used now that any time because of its many advantages. Gamma radiation has high penetrating power, relatively low chemical reactivity and causes small temperature rise. But on the other hand radiosterilization can lead to
Fred C Tenover et al.
International journal of antimicrobial agents, 38(4), 352-354 (2011-08-09)
The activity of ACHN-490 was evaluated against 493 meticillin-resistant Staphylococcus aureus (MRSA) isolates collected in 2009-2010 from 23 US hospitals. The MIC(50) and MIC(90) values (minimal inhibitory concentrations for 50% and 90% of the organisms, respectively) for ACHN-490 were 1
Makoto Adachi et al.
Medical mycology, 45(4), 347-349 (2007-05-19)
Using a newly-developed medium, i.e., combined deactivators-supplemented agar medium (CDSAM), the viability of dermatophytes in skin scales was evaluated. Culture studies were conducted with skin scales collected from 44 patients with tinea pedis who had been treated for two weeks
Yingjian Lu et al.
Marine biotechnology (New York, N.Y.), 11(6), 717-724 (2009-03-12)
Strain GB-2 is a marine microbe with broad-spectrum antimicrobial activity, isolated from soil taken from the coastal city Lianyungang in the JiangSu province of China. Analysis of its morphological, physiological, and biochemical characteristics as well as chemical components of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service