Skip to Content
Merck

Skip To

P136

Putrescine dihydrochloride

Synonym(s):

1,4-Butanediamine dihydrochloride, 1,4-Diaminobutane dihydrochloride, Tetramethylenediamine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NH2(CH2)4NH2 · 2HCl
CAS Number:
Molecular Weight:
161.07
EC Number:
206-375-9
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
3906680
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

280 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].NCCCCN

InChI

1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H

InChI key

XXWCODXIQWIHQN-UHFFFAOYSA-N

Biochem/physiol Actions

Binds to the polyamine modulatory site of the NMDA receptor and potentiates NMDA-induced currents; precursor of spermidine.
Binds to the polyamine modulatory site of the NMDA receptor; potentiates NMDA-induced currents.


Still not finding the right product?

Explore all of our products under Putrescine dihydrochloride


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 1

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library