Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9135-67

Protein A_G High Performance Spintrap Buffer Kit

Cytiva 28-9135-67, sufficient for 16 reactions

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€324.00

€324.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€324.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.25

€324.00


Please contact Customer Service for Availability

sterility

non-sterile

usage

sufficient for 16 reactions

feature

wetted part: no

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9135-67

matrix

highly cross-linked 6% agarose

long term

3-9

short term

2-9

capacity

>10 mg binding capacity(human IgG/ml medium)

Related Categories

General description

The buffers provided in the kit are optimized for standard affinity-based enrichment/immunoprecipitation according to the protocol provided with Protein A HP SpinTrap, Protein G HP SpinTrap, and Ab SpinTrap columns

Features and Benefits

  • Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation using Protein A HP, Protein G HP, and Ab SpinTrap columns.
  • High quality buffers.
  • All buffer components listed and defined.
  • Convenient format - stock solutions can be diluted directly into the provided bottles for storage.
  • Eliminates time-consuming buffer preparation, which allows fast, reproducible protein enrichment.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

SpinTrap is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Hazard Statements

Storage Class Code

8B - Non-combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service