Skip to Content
Merck
All Photos(1)

Key Documents

G8791

Sigma-Aldrich

Gly-Gln solution

200 mM, sterile-filtered, Biotechnology Performance Certified

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

grade

Biotechnology Performance Certified

sterility

sterile-filtered

concentration

200 mM

impurities

endotoxin, tested

storage temp.

2-8°C

SMILES string

NCC(=O)N[C@@H](CCC(N)=O)C(O)=O

Application

Glutamine is an essential component of cell culture media but it is unstable in solution. To address this problem alternatives have been sought. Dipeptides such as gly-L-glutamine and L-ala-L-glutamine have proven to be effective stable forms of glutamine for use in cell culture.

Features and Benefits

Ready-to-use sterile filtered solution

Components

Contains the dipeptide L-Glycyl-L-glutamine. 200 mM supplied in 0.85% NaCL.

comparable product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Minamoto et al.
Cytotechnology, 5 Suppl 2, S35-S51 (1991-01-01)
We tried to establish a new serum-free and heat-sterilizable medium, based on our serum-free medium in which many lymphoblastoid cells and hybridoma could grow as well as in a conventional serum-containing medium. As is well-known, L-glutamine (L-Gln) is one of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service