Skip to Content
Merck
All Photos(1)

Key Documents

F8763

Supelco

Flunitrazepam solution

drug standard, 1.0 mg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12FN3O3
CAS Number:
Molecular Weight:
313.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

drug standard

Assay

≥98%

drug control

Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

CN(C1=CC=C([N+]([O-])=O)C=C1C(C2=CC=CC=C2F)=NC3)C3=O

InChI

1S/C16H12FN3O3/c1-19-14-7-6-10(20(22)23)8-12(14)16(18-9-15(19)21)11-4-2-3-5-13(11)17/h2-8H,9H2,1H3

InChI key

PPTYJKAXVCCBDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Pauly et al.
International clinical psychopharmacology, 26(5), 268-277 (2011-08-11)
Fourteen benzodiazepine (BZD) or BZD-like medications were analyzed with three data sources aiming to assess prescription drug abuse for the year 2008. After a descriptive analysis, a principal component analysis was carried out to explore correlations between seven indicators obtained
T Suzuki et al.
Endoscopy, 44(1), 38-42 (2011-12-07)
Studies have estimated that failure of cecal intubation occurs with conventional colonoscopy in up to 10 % of cases. Double-balloon endoscopy (DBE) systems, magnetic endoscope imaging (MEI), and transparent cap have been shown to improve success rates for colonoscopy. This study
M A Mattila et al.
Drugs, 20(5), 353-374 (1980-11-01)
Flunitrazepam is a benzodiazepine derivative whose hypnotic effect predominates over the sedative, anxiolytic, muscle-relaxing and anticonvulsant effects characteristic of benzodiazepines. Thus, it is used as a night-time hypnotic and in anaesthesiology: due to the pronounced hypnotic effect it is not
Paul D'Aloise et al.
Science & justice : journal of the Forensic Science Society, 52(1), 2-8 (2012-02-14)
Desorption electrospray ionization-mass spectrometry (DESI-MS), a novel ambient ionization technique, was used in this study for determining flunitrazepam in various alcoholic beverages. Using this technique, no pretreatment of the samples was necessary and identification of the drug was accomplished in
Cristina Wasowski et al.
Biochemical pharmacology, 83(2), 253-259 (2011-11-08)
A set of sulfamides designed, synthesized and evaluated against maximal electroshock seizure (MES) and pentilenetetrazol (PTZ) tests with promising results, were tested for their affinity for the benzodiazepine binding site of the GABA(A) receptor. The most active compounds, N,N'-dicyclohexylsulfamide (7)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service