Skip to Content
Merck
All Photos(1)

Key Documents

EMU062011

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ccr7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGTCTTCCAGCTGCCCTACAATGGGGTGGTCCTGGCTCAGACGGTGGCCAACTTCAACATCACCAATAGCAGCTGCGAAACCAGCAAGCAGCTCAACATTGCCTATGACGTCACCTACAGCCTGGCCTCCGTCCGCTGCTGCGTCAACCCTTTCTTGTATGCCTTCATCGGCGTCAAGTTCCGCAGCGACCTCTTCAAGCTCTTCAAGGACTTGGGCTGCCTCAGCCAGGAACGGCTCCGGCACTGGTCTTCCTGCCGGCATGTACGGAACGCGTCGGTGAGCATGGAGGCGGAGACCACCACAACCTTCTCCCCGTAGGGGGCTCCCCTGCCCGGACTACAAGGACCTCTCCCAGGAGCCTTAATGTGGTGCACACATGCACAGACTCTCCATCCACCGAATTG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Li et al.
Medical oncology (Northwood, London, England), 31(9), 180-180 (2014-08-22)
Secondary lymphoid tissue chemokine (SLC/CCL21) and its receptor CCR7 have been implicated in lymph node metastasis, whereas the mechanism of which remains unclear. Epithelial-mesenchymal transition (EMT) plays an important role in invasion and migration of cancer cells. We presumed that
Kaori Kubo et al.
Biochemical and biophysical research communications, 463(4), 825-831 (2015-06-24)
Chronic myeloid leukemia is a clonal disease characterized by the presence of the Philadelphia chromosome and its oncogenic product, BCR-ABL, which activates multiple pathways involved in cell survival, growth promotion, and disease progression. We previously reported that in murine hematopoietic
Yang Yue et al.
Oncology reports, 34(6), 3280-3287 (2015-09-10)
In the present study, we aimed to demonstrate whether praline-rich tyrosine kinase-2 (Pyk2) participates in the chemokine receptor 7 (CCR7) downstream signaling network, and to determine the role of this molecule and the related mechanism in the CCR7-mediated regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service