Skip to Content
Merck

Skip To

D4159

cis-13,16-Docosadienoic acid

~99% (capillary GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H40O2
CAS Number:
Molecular Weight:
336.55
UNSPSC Code:
12352211
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~99% (capillary GC)

storage temp.

−20°C

SMILES string

CCCCC\C=C\C\C=C\CCCCCCCCCCCC(O)=O



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J M Muderhwa et al.
The Journal of biological chemistry, 267(34), 24184-24192 (1992-12-05)
Pancreatic carboxylester lipase catalyzes the exchange of 18O between water and 13,16-cis,cis-doco-sadienoic acid (DA) in monolayers at the argon-buffer interface (Muderhwa, J.M., Schmid, P.C., and Brockman, H.L. (1992) Biochemistry 31, 141). In mixed monolayers of 18O, 18O-DA and 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC)
G D Schmit et al.
Biophysical journal, 71(6), 3421-3429 (1996-12-01)
It has been suggested that at physiological pH, the trypsin-catalyzed activation of the lipase cofactor, procolipase, to colipase has no consequence for intestinal lipolysis and serves primarily to release the N-terminal pentapeptide, enterostatin, a satiety factor (Larsson, A., and C.
Claire Denis et al.
Lipids, 44(7), 655-663 (2009-03-07)
The phospholipid fatty acid composition of the North-East Atlantic sponge Polymastia penicillus (South Brittany, France) was investigated. Sixty fatty acids (FA) were identified as methyl esters (FAME) and N-acyl pyrrolidides (NAP) by gas chromatography-mass spectrometry (GC/MS), including eight Delta5,9 unsaturated