Skip to Content
Merck
All Photos(1)

Documents

CRISPR13

Sigma-Aldrich

CRISPR-Lenti Human HPRT1 Positive Control DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NE.02

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

Related Categories

Application

Functional Genomics/Screening/Target Validation
  • Positive control, HPRT1, CRISPR lentivirus DNA format, for creating gene knockouts/genetic modifications in cell lines.

To see more application data, protocols, vector maps visit Sigma.com/crispr.

Features and Benefits

Serves as a positive experimental control for the CRISPR editing workflow using lenti CRISPR DNA. The number of cells with HPRT1 knocked out increases over time with 6−TG selection.

Physical form

The Sigma CRISPR Lenti Human HPRT1 Positive Control Vector is provided as 1 μg of plasmid DNA in Tris−EDTA (TE).

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service