Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
41106514
Biological source:
Mouse x Mouse Hybridoma
Isotype:
IgG1
Technique(s):
cell culture | mammalian: suitable
Myeloma:
NS-1 (P3-X63/Ag8-NS1)
Reactivity:
Anti-Leishmania (L) pifanoi Amastigote
Pricing and availability is not currently available.
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 08041723-DNA-5UG, pkg of vial of cells 08041723-1VL
ImmunoDon
Balb/c mouse spleen
Myeloma
NS-1 (P3-X63/Ag8-NS1)
Reactivity
Anti-Leishmania (L) pifanoi Amastigote
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Immunogen
Membrane from Leishmania (L) pifanoi strain LTRod-MHOM/VE/60/LtRod-Cl-A-3
Application
Detection of Leishmania (L) pifanoi
Biochem/physiol Actions
Mouse NS1 x BALB/c mouse
XCIII-1F8-D7 was developed by immunizing mice with membranes from Leishmania (L) pifanoi and fusing the mouse spleen cells with NS-1(P3-X63/Ag8-NS1)
Preparation Note
RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Seed cultures between 1-3x100,000 cells/ml; Saturation density 1,000,000 5% CO2; 37°C
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to [email protected].
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.