Skip to Content
Merck

Skip To

B6430

Nalgene® dropper bottle with screw closure

capacity 4 mL, white bottle, white cap

Synonym(s):

bottle dropper, dropper bottles, Dropper container, Dropper flask, Nalgene 2751-9125, dispensing bottle

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 ea
Please contact Customer Service for Availability
€62.80

About This Item

NACRES:
NB.17
UNSPSC Code:
41121800

€62.80


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

low-density polyethylene , round bottle, white bottle

joint

Joints threaded neck (15-415)

packaging

pack of 25 ea

manufacturer/tradename

Nalgene® 2751-9125

capacity

4 mL

color

white cap

Features and Benefits

  • Nalgene® dropper bottles with screw closure is an alternative to various pipetting devices.
  • The tip ensures accurate dispensing of 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time.
  • Chemically resistant to most reagents.
  • LDPE clear transparent bottles for easy sample visualization.
  • Leak-proof closure - prevents sample loss.
  • Control dispensing tip ensures a secure fit.
  • Suitable for use in biotech, diagnostic and pharmaceutical applications.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B6805B6055B7055
capacity

4 mL

capacity

4 mL

capacity

4 mL

capacity

15 mL

manufacturer/tradename

Nalgene® 2751-9125

manufacturer/tradename

Nalgene® 2752-9125

manufacturer/tradename

Nalgene® 2750-9125

manufacturer/tradename

Nalgene® 2752-9050

material

low-density polyethylene , white bottle, round bottle

material

low-density polyethylene , round bottle, white bottle

material

colorless bottle, low-density polyethylene , polypropylene bottle, round bottle

material

low-density polyethylene , natural bottle, round bottle

joint

Joints threaded neck (15-415)

joint

Joints threaded neck (15-415)

joint

Joints threaded neck (15)

joint

Joints threaded neck (15)

packaging

pack of 25 ea

packaging

pack of 25 ea

packaging

pack of 25 ea

packaging

pack of 25 ea

color

white cap

color

assorted colors cap

color

white cap

color

assorted colors cap


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
B6430-25EA04061832613680