33015-M
Formic acid
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
Quality Level
Agency
reag. Ph. Eur.
vapor density
1.6 (vs air)
vapor pressure
44.8 mmHg ( 20 °C)
Assay
≥98%
form
liquid
autoignition temp.
1004 °F
expl. lim.
57 %
technique(s)
mass spectrometry (MS): suitable
impurities
≤0.001% ammonium (NH4)
≤0.001% non-volatile matter
≤0.005% acetic acid (CH3COOH)
refractive index
n20/D 1.370 (lit.)
pH
2.2 (20 °C, 2.2 g/L)
bp
100-101 °C (lit.)
mp
8.2-8.4 °C (lit.)
density
1.22 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤5 mg/kg
oxalate (C2O42-): ≤50 mg/kg
sulfate (SO42-): ≤5 mg/kg
sulfite (SO32-): ≤10 mg/kg
cation traces
Ag: ≤0.02 mg/kg
Al: ≤0.05 mg/kg
Ba: ≤0.05 mg/kg
Be: ≤0.02 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.2 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.05 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤2 mg/kg
Ge: ≤0.05 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.02 mg/kg
Mg: ≤0.5 mg/kg
Mn: ≤0.05 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.05 mg/kg
Pb: ≤0.02 mg/kg
Sr: ≤0.02 mg/kg
Ti: ≤0.1 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.05 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.1 mg/kg
SMILES string
OC=O
InChI
1S/CH2O2/c2-1-3/h1H,(H,2,3)
InChI key
BDAGIHXWWSANSR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
121.1 °F - closed cup
Flash Point(C)
49.5 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service