Skip to Content
Merck
All Photos(1)

Documents

27001-M

Sigma-Aldrich

Formic acid

puriss., meets analytical specifications of DAC, FCC, 98.0-100%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

1.6 (vs air)

Quality Level

vapor pressure

44.8 mmHg ( 20 °C)

grade

puriss.

Assay

98.0-100%

form

liquid

autoignition temp.

1004 °F

quality

meets analytical specifications of DAC, FCC

expl. lim.

57 %

impurities

≤0.001% heavy metals (as Pb)
≤0.05% acetic acid (GC)

evapn. residue

≤0.005%

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 2.2 g/L)

bp

100-101 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 ppm
sulfate (SO42-): ≤40 ppm
sulfite (SO32-): in accordance

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Determination of Cotinine and 3-hydroxynicotinine in Human Serum: This study presents a method for detecting nicotine metabolites in human serum using liquid chromatography-tandem mass spectrometry, where formic acid is used to aid in the extraction and detection process (Wang et al., 2024).
  • Quantification of Antiretrovirals in Human Tissues: Focuses on a method for quantifying antiretrovirals in tissues using ultra-high performance liquid chromatography-tandem mass spectrometry, where formic acid may enhance the analysis (West et al., 2024).
  • Redox Regulation of Ni Hydroxides: This research into the electro-refinery of biomass-derived polyols may involve formic acid in the preparation of samples or in the electrolyte solutions used (Gui et al., 2024).
  • Volatile Fatty Acids in Biological Specimens: Discusses a technique for determining volatile fatty acids in various biological samples using HPLC-MS/MS with a low-temperature derivatisation method that likely involves formic acid (Li et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Athar Masood et al.
Basic & clinical pharmacology & toxicology, 127(6), 533-550 (2020-11-03)
Urine drug testing (UDT) is an important analytical/bio-analytical technique that has inevitably become an integral and vital part of a testing programme for diagnostic purposes. This manuscript presents a tailor-made LC-MS/MS quantitative assay method development and validation for a custom
Heng Lu et al.
Cell communication insights, 5, 1-7 (2014-05-20)
Protein-protein interactions play a pivotal role in both inter- and intra-cellular signaling. Identification of signaling protein complexes can thus shed important new insights into cell communications. We developed a parallel affinity precipitation protocol to overcome the disadvantages of the tandem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service