Skip to Content
Merck
All Photos(1)

Documents

04316

Millipore

Vegetable Extract No. 1

suitable for microbiology

Synonym(s):

Meat Extract Replacement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.85

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥3%
total nitrogen (N), ≥9%

impurities

≤12% ash

loss

≤6% loss on drying

pH

6.6±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, slightly yellow

application(s)

food and beverages
vitamins, nutraceuticals, and natural products

microbiology

General description

Vegetable Extract No. 1 is a meat-free alternative to traditional peptones. With a wide distribution of peptides it meets the requirements for replacing Meat Extract (Cat. No. 70164) to yield maximum growth of fastidious microorganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service