SPKGENPNE001
Spare part, Filter Cartridge
40µm Filtration grade
Select a Size
€598.00
In StockDetails
Select a Size
About This Item
€598.00
In StockDetails
General description
This spare part is required for the preventive maintenance service of some of our BioProcessing systems, it is recommended to have one in stock and to change it every 12 months.