Key Documents
SPKGENELC045
Millipore
Spare part, filter mat
W × H × D 173 mm × 173 mm × 17 mm
Select a Size
SPKGENELC045
€829.00
€829.00
Please contact Customer Service for Availability
Select a Size
Change View
SPKGENELC045
€829.00
About This Item
UNSPSC Code:
23151806
€829.00
Please contact Customer Service for Availability
material
fibrous
Quality Level
product line
Mobius®
W × H × D
173 mm × 173 mm × 17 mm
compatibility
for use with Mobius®
General description
These filters are used on BioProcessing systems to capture dust, dirt, and other airborne particles. They ensure optimal performance and of the equipment.
This spare part is required for the preventive maintenance service of some of our BioProcessing systems, it is recommended to have one in stock and to change it every 12 months.
Packaging
Pack of 5 units
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany