Skip to Content
Merck
All Photos(2)

Key Documents

SCT083

Sigma-Aldrich

BioTracker MemBright 488 Live Cell Dye

Synonym(s):

Live cell imaging probe, Live cell membrane probe, green

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€183.00

€183.00


Estimated to ship onApril 08, 2025Details



Select a Size

Change View
1 VIAL
€183.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.47

€183.00


Estimated to ship onApril 08, 2025Details


mol wt

observed mol wt 1218.5 kDa

Quality Level

packaging

vial of 50 μL

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell based assay: suitable
flow cytometry: suitable

detection method

fluorometric

shipped in

dry ice

storage temp.

−20°C

General description

The BioTracker Membright-488 live cell dye is a “turn-on” fluorescence probe emitting in the far red spectrum for the imaging of plasma membranes of live cells.

Specific plasma membrane markers are essential for delineating the cell. Membright dyes exploit a unique fluorogenic mechanism, existing in a self-quenched nanoparticle state. Fluorescence is turned on upon contact with and propagation within the plasma membrane, producing bright, background-free signal. The Membright live cell membrane dyes are compatible with two-photon imaging, superresolution imaging, and fixed cell preparations. The high photostability and lack of cytotoxicity of Membright dyes make them suitable for long-term live cell imaging. The suite of superior characteristics of Membright plasma membrane dyes permit use in a wide spectrum of fluorescence microscopy applications and flow cytometry.

Spectral Properties
Fluorescence images obtained by λex = 488 nm and emission range of 500-550 nm.

Reference

Collot M et al. MemBright: A family of fluorescent membrane probes for advanced cellular imaging and neuroscience. Cell Chem. Biol. 2019, 26:600-614

Application

Purity: > 98% confirmed by HPLC, HNMR, LC-MS and elemental analysis.

Features and Benefits

Green fluorescent probe for efficient, selective labeling of the plasma membrane in live cells. Suitable for superresolution microscopy.

Physical form

Green solution

Storage and Stability

Store long term at -20°C, protected from light.

Other Notes

Live cell fluorescent imaging, flow cytometry

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the concentration of membright 488. Only info I have is that it is 50 microlitres. Publications use it at 20 nM

    1 answer
    1. The concentration provided at 200uM in DMSO solution, final volume 50ul. Please spin down this material before usage to be able to collect all the solution in the bottom of the tube. It is recommended to use the material at 1:1000 dilution, but this is sample dependent.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service