JX-1L-BK
Intermediates for Further Manufacturing Use (FFMU) - RHCE (Rh) Specificities
Synonym(s):
Anti-e (clone MS-69), FFMU
About This Item
Recommended Products
pH
6.7-7.7
Quality Level
Related Categories
General description
For the formulation of Anti-e blood typing reagents or devices.
Reactivity is greatly enhanced by chemical potentiation; Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.
Application
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service