Key Documents
XXSPTFF02
Millipore
Pellicon® Single-Pass TFF Cassette Diverter Plate
Select a Size
XXSPTFF02
€1,773.00
€1,773.00
Estimated to ship onJuly 28, 2025Details
Select a Size
Change View
XXSPTFF02
€1,773.00
About This Item
UNSPSC Code:
23151800
eCl@ss:
32031690
€1,773.00
Estimated to ship onJuly 28, 2025Details
material
316L stainless steel housing
Quality Level
technique(s)
tangential flow filtration (TFF): suitable
application(s)
liquid formulation
General description
The Pellicon Single-Pass TFF Cassette Diverter Plate is used to configure Pellicon 3 0.57 m², Pellicon 3 1.14 m² or Pellicon 2 0.5 m² cassettes for single-pass TFF operation.
Legal Information
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany