Skip to Content
Merck
All Photos(2)

Key Documents

ABN1441

Sigma-Aldrich

Anti-ABCA4

from rabbit, purified by affinity chromatography

Synonym(s):

Retinal-specific ATP-binding cassette transporter, ATP-binding cassette sub-family A member 4, RIM ABC transporter, RIM protein, RmP, Stargardt disease protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€371.00

€371.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€371.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€371.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ABCA4(24)

General description

Retinal-specific ATP-binding cassette transporter (UniProt: P78363; also known as ATP-binding cassette sub-family A member 4, RIM ABC transporter, RIM protein, RmP, Stargardt disease protein) is encoded by the ABCA4 (also known as ABCR) gene (Gene ID: 24) in human. ABC transporter A4 is a multi-pass membrane protein of the superfamily of ATP-binding cassette (ABC) transporters that is localized to the disk margins of vertebrate photoreceptor outer segments. It is implicated in the clearance of all-trans retinal derivatives from rod and cone photoreceptor cells following photoexcitation as part of the visual cycle. ABC transporter A4 is composed of two symmetrically organized halves, each comprising six membrane-spanning helices, a large glycosylated exocytoplasmic domain located inside the disk, and a cytoplasmic domain with an ATP-binding cassette. Mutations in ABCA4 gene are known to cause Stargardt macular degeneration, which is an autosomal recessive form of juvenile macular degeneration associated with a severe loss in vision. Mutations in ABCA4 gene are also known to cause retinitis pigmentosa, which is characterized by retinal pigment deposits and loss of rod photoreceptor cells followed by secondary loss of cone photoreceptors. (Ref.: Tsybovsky, Y., et al. (2010). Adv. Exp. Med. Biol. 703, 105-125).

Specificity

This rabbit polyclonall antibody detects Retinal-specific ATP-binding cassette transporter (ABCA4) in Human retina. It targets an epitope within 112 amino acids from the cytoplasmic domain.

Immunogen

GST-tagged reombinant fragment corresponding to 112 amino acids from the internal region of human Retinal-specific ATP-binding cassette transporter (ABCA4).

Application

Anti-ABCA4, Cat. No. ABN1441, is a highly specific rabbit polyclonal antibody that targets Retinal-specific ATP-binding cassette transporter (ABCA4) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected ABCA4 in human retina tissue.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human retina tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected ABCA4 in 10 µg of human retina tissue lysate.

Target description

~256 kDa observed; 255.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service