Skip to Content
Merck
All Photos(2)

Key Documents

ABN1469

Sigma-Aldrich

Anti-Alpha-2-HS-Glycoprotein

from rabbit, purified by affinity chromatography

Synonym(s):

Alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, Fetuin-A, Alpha-2 Heremans-Schmid glycoprotein, AHGS

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€151.00
100 μL
€415.00

€151.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€151.00
100 μL
€415.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€151.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... AHSG(197)

General description

Alpha-2-HS-glycoprotein (UniProt: P02765; also known as Alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, Fetuin-A, Alpha-2 Heremans-Schmid glycoprotein, AHSG) is encoded by the AHSG (also known as FETUA, PRO2743) gene (Gene ID: 197) in human. Alpha-2-HS-glycoprotein is a serum protein of the cystatin superfamily that is synthesized by hepatocytes and is considered as a negative acute-phase reactant. It is highly expressed in various fetal tissues, but in adults it is mainly expressed in the liver. It is synthesized with a signal peptide (aa 1-18) and a connective propeptide (aa 301-340) and is subsequently cleaved into 2 chains known as Alpha-2-HS-glycoprotein chain A and Alpha-2-HS-glycoprotein chain B. The mature product is involved in multiple normal biological and pathological functions. It promotes possesses opsonic properties and is shown to be involved in endocytosis, brain development, bone resorption, regulation of insulin activity and hepatocyte growth factor (HGF) activity. Alpha-2-HS-glycoprotein also serves as a carrier protein and serves as a carrier for otherwise insoluble calcium phosphate and can act as a potent inhibitor of pathological vascular calcification. Higher levels of Alpha-2-HS-glycoprotein have been reported in gestational and type II diabetes. It is also present at higher levels in patients with endometriosis. Lower levels of Alpha-2-HS-glycoprotein are present in cancer subjects. (Ref.: Kalabay, L., et al. (2002). Eur. J. Endocrinol. 147(2); 243-248; Sindhu, S., et al. (2016). BMC Immunol. 17; 33).

Specificity

This rabbit polyclonal antibody specifically detects human Alpha-2-HS-glycoprotein. It targets an epitope with in the N-terminal half.

Immunogen

Epitope: N-terminus
GST/His-tagged recombinant fragment corresponding to 146 amino acids from human Alpha-2-HS-glycoprotein.

Application

Anti-Alpha-2-HS-Glycoprotein Antibody, Cat. No. ABN1469, is a highly specific rabbit polyclonal antibody that targets Alpha-2-HS-Glycoprotein and has been tested for use Immunohistochemistry and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A 1:500 dilution from a representative lot detected Alpha-2-HS-Glycoprotein in human retina tissue lysate.

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Alpha-2-HS-Glycoprotein in human liver and human bone marrow tissue sections.

Quality

Evaluated by Western Blotting in human serum.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Alpha-2-HS-Glycoprotein in human serum.

Target description

~55 kDa observed; 39.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service