Skip to Content
Merck

Skip To

ABE441

Anti-dimethyl Histone H4 (Arg3), Asymmetric Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H4

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
DB, ICC, WB
Citations:
-


biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

dot blot: suitable, immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Arg3)

Gene Information

human ... HIST2H4B(554313)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The histones have an amino terminal tail, a globular domain, and a carboxy-terminal tail. The four core histones, H2A, H2B, H3 and H4 assemble into the octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped in a sequence-independent manner around the octamer, forming the basic subunit of chromatin, the nucleosome. Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.
~13 kDa observed

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4, asymmetrically dimethylated at Arg3.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected dimethyl Histone H4 (Arg3), Asymmetric in HeLa cells.

Dot Blot (Specificity) Analysis: A 1:10,000 dilution from a representative lot detected dimethyl Histone H4 (Arg3), Asymmetric in an Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and an Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).

DyLight is a registered trademark of Thermo Fisher Scientific.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-dimethyl Histone H4 (Arg3) antibody, Asymmetric is validated for use in western blotting, ICC & dot blot for the detection of dimethyl Histone H4 (Arg3).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in untreated and CARM1 treated recombinant Histone H4.

Western Blotting Analysis: A 1:50,000 dilution of this antibody detected dimethyl Histone H4 (Arg3), Asymmetric in 0.5 µg of CARM1 treated recombinant Histone H4.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABE2909SAB5700195SAB5700210
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

UniProt accession no.

P62805

UniProt accession no.

P62805

UniProt accession no.

P62805

UniProt accession no.

P62805

shipped in

wet ice

shipped in

ambient

shipped in

wet ice

shipped in

wet ice

species reactivity

human

species reactivity

human

species reactivity

human, rat, mouse

species reactivity

mouse, human, rat

technique(s)

dot blot: suitable, western blot: suitable, immunocytochemistry: suitable

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, western blot: suitable

technique(s)

immunofluorescence: 1:50-1:200, immunohistochemistry: 1:50-1:200, western blot: 1:500-1:2000

technique(s)

ChIP: 1:50-1:200, immunofluorescence: 1:50-1:200, western blot: 1:500-1:2000


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Cancer is a complex disease manifestation. At its core, it remains a disease of abnormal cellular proliferation and inappropriate gene expression. In the early days, carcinogenesis was viewed simply as resulting from a collection of genetic mutations that altered the gene expression of key oncogenic genes or tumor suppressor genes leading to uncontrolled growth and disease (Virani, S et al 2012). Today, however, research is showing that carcinogenesis results from the successive accumulation of heritable genetic and epigenetic changes. Moreover, the success in how we predict, treat and overcome cancer will likely involve not only understanding the consequences of direct genetic changes that can cause cancer, but also how the epigenetic and environmental changes cause cancer (Johnson C et al 2015; Waldmann T et al 2013). Epigenetics is the study of heritable gene expression as it relates to changes in DNA structure that are not tied to changes in DNA sequence but, instead, are tied to how the nucleic acid material is read or processed via the myriad of protein-protein, protein-nucleic acid, and nucleic acid-nucleic acid interactions that ultimately manifest themselves into a specific expression phenotype (Ngai SC et al 2012, Johnson C et al 2015). This review will discuss some of the principal aspects of epigenetic research and how they relate to our current understanding of carcinogenesis. Because epigenetics affects phenotype and changes in epigenetics are thought to be key to environmental adaptability and thus may in fact be reversed or manipulated, understanding the integration of experimental and epidemiologic science surrounding cancer and its many manifestations should lead to more effective cancer prognostics as well as treatments (Virani S et al 2012).






Questions

Reviews

No rating value

Active Filters