Skip to Content
Merck
All Photos(2)

Key Documents

ABE30

Sigma-Aldrich

Anti-RNA polymerase II subunit B1 Antibody, clone 4F8

from rabbit, purified by affinity chromatography

Synonym(s):

polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, DNA-directed RNA polymerase III largest subunit, DNA-directed RNA polymerase II subunit RPB1, RNA polymerase II subunit B1, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€354.00

€354.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€354.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€354.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), horse (based on 100% sequence homology), canine (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POLR2B(5431)

General description

RNA polymerase II subunit B1 (RPB1) is the largest subunit of the RNA polymerase II complex. As a holoenzyme RNA polymerase II catalyzes transcription of eukaryotic DNA into RNA using the four ribonucleoside triphosphates as substrates. The RPB1 subunit, in combination with other polymerase subunits, forms a large central cleft that maintains contact between the active site of the enzyme, the DNA template, and the nascent RNA transcript. This subunit also contains a carboxy terminal domain (CTD) consisting of tandem heptapeptide repeats. In actively transcribing RNA polymerase ‘Ser-2’ and ‘Ser-5’ of the heptapeptide repeat are phosphorylated. Phosphorylation activates the RNA polymerase II beta subunit, allowing it to serve as an assembly platform for additional subunits that modulate initiation, elongation, termination and mRNA processing.

Immunogen

KLH-conjugated linear peptide corresponding to human RNA polymerase II subunit B1.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected RNA polymerase II subunit B1 in HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-RNA polymerase II subunit B1 Antibody, clone 4F8 (rabbit polyclonal antibody) validated in WB, ICC to detect RNA polymerase II subunit B1 also known as DNA-directed RNA polymerase II subunit RPB1.

Quality

Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 1 µg/mL of this antibody detected RNA polymerase II subunit B1 on 10 µg of HeLa nuclear extract.

Target description

~217 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced RAN expression and disrupted transport between cytoplasm and nucleus; a key event in Alzheimer's disease pathophysiology.
Mastroeni, D; Chouliaras, L; Grover, A; Liang, WS; Hauns, K; Rogers, J; Coleman, PD
Testing null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service