Skip to Content
Merck
All Photos(1)

Key Documents

AB5761

Sigma-Aldrich

Anti-Noxa Antibody, a.a. 1-16

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€370.00

€370.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μL
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PMAIP1(5366)

Specificity

Noxa. Western blot analysis of cells infected with Noxa adenoviruses and BAF indicates a high level of specificity for the 16.5 kDa Noxa protein.

Immunogen

Epitope: a.a. 1-16
Synthetic peptide from amino acids 1-16 of mouse Noxa. 15/16 amino acids in this sequence are conserved in rat and 10/16 conserved in human.

Application

Research Category
Neuroscience
Research Sub Category
Oxidative Stress
This Anti-Noxa Antibody, a.a. 1-16 is validated for use in WB for the detection of Noxa.
Western blot: 1:1,000-1:2,000

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50 μL of sterile distilled water.

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preedakorn Chunhacha et al.
Cell death discovery, 7(1), 310-310 (2021-10-25)
Heart failure (HF) has reached epidemic proportions in developed countries, affecting over 20 million people worldwide. Despite modern medical and device therapies, 60-70% of HF patients still die within 5 years of diagnosis as it relentlessly progresses through pervasive apoptotic
Xue Gong et al.
The Journal of clinical investigation, 131(24) (2021-12-16)
Circular RNAs (circRNAs) have been recently recognized as playing a role in the pathogenesis of vascular remodeling-related diseases by modulating the functions of miRNAs. However, the interplay between circRNAs and proteins during vascular remodeling remains poorly understood. Here, we investigated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service