Merck
  • Fortilin inhibits p53, halts cardiomyocyte apoptosis, and protects the heart against heart failure.
Fortilin inhibits p53, halts cardiomyocyte apoptosis, and protects the heart against heart failure.
Cell death discovery (2021-10-25)
Preedakorn Chunhacha, Decha Pinkaew, Patuma Sinthujaroen, Dawn E Bowles, Ken Fujise
ABSTRACT
Heart failure (HF) has reached epidemic proportions in developed countries, affecting over 20 million people worldwide. Despite modern medical and device therapies, 60-70% of HF patients still die within 5 years of diagnosis as it relentlessly progresses through pervasive apoptotic loss of cardiomyocytes. Although fortilin, a 172-amino-acid anti-p53 molecule, is one of the most expressed proteins in the heart, its precise role there has remained unknown. Also unclear is how cardiomyocytes are protected against apoptosis. Here, we report that failing human hearts express less fortilin than do non-failing hearts. We also found that mice lacking fortilin in the heart (fortilinKO-heart) die by 9 weeks of age due to extensive cardiomyocyte apoptosis and severe HF, which suggests that fortilin sustains cardiomyocyte viability. The lack of fortilin is also associated with drastic upregulation of p53 target genes in the hearts. The heart-specific deletion of p53 in fortilinKO-heart mice extends their life spans from 9 to 18 weeks by mitigating cardiomyocyte apoptosis. Our data suggest that fortilin is a novel cardiac p53 inhibitor and that its inadequate expression in failing hearts and subsequent overactivation of the p53 apoptosis pathway in cardiomyocytes exacerbates HF.
Sigma-Aldrich
Anti-Noxa Antibody, a.a. 1-16, serum, Chemicon®