Skip to Content
Merck

Skip To

AB5482

Anti-Superoxide Dismutase 1 Antibody

serum, Chemicon®

Synonym(s):

SOD1

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
6


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

should not react with

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SOD1(6647)

General description

Mutations in the copper/ zinc superoxide dismutase (SOD1) gene are associated with 15- 20% of the familial forms of motoneuron disease. This 153 amino acid metalloenzyme is expressed in virtually all cells of all organisms above bacteria and is highly conserved across species, although some minor variations do occur. The incorporation of the mutated form of the SOD1 human gene into a transgenic mouse leads to the onset of the disease that closely resembles the human condition. These animals become weak at about 2-4 months of age and rapidly lose function, which results in death 4- 6 weeks later.

Immunogen

Synthetic peptide corresponding to amino acids 25-37 of mouse SOD1.

Application

Detect Superoxide Dismutase 1 using this Anti-Superoxide Dismutase 1 Antibody validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress
Western blotting: 1:200-1:500 (see images)

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

Superoxide Dismutase 1 (SOD1), mouse and rat specific. No reactivity with human SOD1

Physical form

Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50 μL of sterile water.

Preparation Note

Maintain lyophilized material at -70°C (dry) for up to 12 months after date of receipt. After reconstitution maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Glycerol (1:1) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB5480SAB1406464S2147
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

antibody form

serum

antibody form

serum

antibody form

purified immunoglobulin

antibody form

ascites fluid

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

rat, mouse

species reactivity

human

species reactivity

human

species reactivity

canine, human, rat

Gene Information

human ... SOD1(6647)

Gene Information

human ... SOD1(6647)

Gene Information

human ... SOD1(6647)

Gene Information

human ... SOD1(6647)
rat ... Sod1(24786)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters