Skip to Content
Merck
All Photos(2)

Key Documents

529600

Sigma-Aldrich

Prionex® Reagent

A unique alternative to bovine serum albumin (BSA).

Synonym(s):

Protein stabilizer and blocking agent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤10 EU/mL Endotoxins

color

clear pale yellow

cation traces

heavy metals: ≤20 ppm

shipped in

ambient

storage temp.

2-8°C

General description

A unique alternative to bovine serum albumin (BSA). Prionex® Reagent is a polypeptide fraction of highly purified dermal collagen of porcine origin that is free from cartilage, bone and plasma components and that can be considered as an extremely pure form of gelatin type A. Used as an inert protein stabilizer in many applications. Also useful as a blocking agent and as a protective additive in cell culture.

Warning

Toxicity: Standard Handling (A)

Physical form

Sterile aqueous solution pH 6.0-7.5.

Reconstitution

Following initial use, aliquot and freeze (-20°C) for long-term storage.

Other Notes

Gaffney, P.J., et al. 1996. J. Pharm. Pharmacol.48, 896.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Prionex is a registered trademark of Pentapharm AG, Basel

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service