1468500006
Tryptone Azolectin (TAT) Broth
bottle of 190 mL
Synonym(s):
Casein Lecithin Tween Broth, Casein Peptone Lecithin Polysorbate, TAT Broth, Tryptic Soy Lecithin Polysorbate Broth, Tryptic Soya Lecithin Polysorbate Broth
About This Item
Recommended Products
Agency
EP
JP 4.06
USP
sterility
sterile; autoclaved
form
liquid
feature
ready-to-use
packaging
bottle of 190 mL
pH
7.1±0.2 (25 °C)
application(s)
cosmetics
food and beverages
pharmaceutical
sterility testing
storage temp.
2-25°C
suitability
nonselective for
General description
Application
Features and Benefits
Legal Information
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service