Skip to Content
Merck
All Photos(1)

Key Documents

1.46495

Millipore

Casein Peptone Lecithin Polysorbate Broth

ready-to-use, bottle volume 90 mL , filling volume

Synonym(s):

Casein Peptone Lecithin Polysorbate, TAT Broth, Tryptic Soy Lecithin Polysorbate Broth, Tryptone Soya Lecithin Polysorbate Broth, Casein Lecithin Tween Broth

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 90 ML
€46.10

€46.10


Please contact Customer Service for Availability


Select a Size

Change View
10 X 90 ML
€46.10

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

€46.10


Please contact Customer Service for Availability

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

description

Suitable for diluting pharmaceutical and cosmetic products for microbial count determination

sterility

sterile; autoclaved

form

liquid (ready-to-use)

feature

closure type (screw cap with septum)
ready-to-use

packaging

pkg of 90 mL (in 125 ml bottle with blue screw cap and 3 loci (10 bottles per box))
bottle of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

125 mL

bottle volume

90 mL , filling volume

pH

7.1

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

components

tween
lecithin

storage temp.

2-25°C

suitability

nonselective for

General description

Casein Peptone Lecithin Polysorbate Broth contains relatively high casein peptone that provides optimal conditions for the spore germination and regeneration of microorganisms. The neutralizing additives reverse the growth inhibitory effect of most disinfectants and antiseptics. Lecithin from soya inactivates cetrimide, chlorohexidine, chlorinated phenols, desqualinium acetate, and polymyxin B. Polysorbate (Tween®) 20 inactivates phenols, phenol derivates, benzoic acid, p-hydroxybenzoic acid, and their esters. The combination of lecithin and polysorbate (Tween®) 20 inactivates many antimicrobial compounds such as quaternary ammonium and phosphonium compounds.

Application

Casein Peptone Lecithin Polysorbate Broth (TAT Broth) is used for diluting samples of pharmaceutical, cosmetic and other raw materials or final products when determining microbial counts.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Our media are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service