Skip to Content
Merck
All Photos(1)

Documents

78725

Sigma-Aldrich

O-Phenylhydroxylamine hydrochloride

≥97.0% (AT)

Synonym(s):

Phenoxyamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5ONH2 · HCl
CAS Number:
Molecular Weight:
145.59
Beilstein:
4326059
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (AT)

form

powder

mp

~132 °C (dec.)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cl.NOc1ccccc1

InChI

1S/C6H7NO.ClH/c7-8-6-4-2-1-3-5-6;/h1-5H,7H2;1H

InChI key

DBTXKJJSFWZJNS-UHFFFAOYSA-N

Related Categories

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service