Skip to Content
Merck
All Photos(1)

Key Documents

718157

Sigma-Aldrich

Silver(II) picolinate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8AgN2O4
CAS Number:
Molecular Weight:
352.07
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reagent type: oxidant

mp

172-173 °C

SMILES string

O=C(O[Ag]OC(=O)c1ccccn1)c2ccccn2

InChI

1S/2C6H5NO2.Ag/c2*8-6(9)5-3-1-2-4-7-5;/h2*1-4H,(H,8,9);/q;;+2/p-2

InChI key

WWTBYVQANMUFPG-UHFFFAOYSA-L

Application

Reagent for:
  • Synthesis of the polycyclic dimeric pyrrole-imidazole alkaloid (±)-palau′amine
  • Oxidation reactions (oxidizing agent)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total syntheses of (+/-)-massadine and massadine chloride.
Shun Su et al.
Journal of the American Chemical Society, 130(49), 16490-16491 (2008-12-04)
Total synthesis of palau'amine.
Ian B Seiple et al.
Angewandte Chemie (International ed. in English), 49(6), 1095-1098 (2009-12-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service