Skip to Content
Merck
All Photos(1)

Key Documents

676454

Sigma-Aldrich

Potassium palmitate-1,3,5,7,9-13C5

endotoxin tested, 99 atom % 13C, 98% (CP)

Synonym(s):

Hexadecanoic acid-1,3,5,7,9-13C5 potassium salt, Palmitic Acid-1,3,5,7,9-13C5 potassium salt, Potassium hexadecanoate-1,3,5,7,9-13C5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6(13CH2CH2)413CO2K
CAS Number:
Molecular Weight:
299.48
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

mp

285 °C (dec.)

suitability

endotoxin tested

mass shift

M+5

SMILES string

[K+].CCCCCCC[13CH2]C[13CH2]C[13CH2]C[13CH2]C[13C]([O-])=O

InChI

1S/C16H32O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1/i8+1,10+1,12+1,14+1,16+1;

InChI key

MQOCIYICOGDBSG-DVEYMGBSSA-M

Related Categories

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.

Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Citi et al.
Pharmacological research, 159, 105039-105039 (2020-06-23)
The gasotransmitter hydrogen sulfide (H2S) is involved in the regulation of the vascular tone and an impairment of its endogenous production may play a role in hypertension. Thus, the administration of exogenous H2S may be a possible novel and effective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service