Skip to Content
Merck
All Photos(1)

Key Documents

333034

Sigma-Aldrich

Tributyltin iodide

technical grade, 90%

Synonym(s):

Iodotributyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnI
CAS Number:
Molecular Weight:
416.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical grade

Assay

90%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.53 (lit.)

bp

172 °C/10 mmHg (lit.)

density

1.46 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](I)(CCCC)CCCC

InChI

1S/3C4H9.HI.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;1H;/q;;;;+1/p-1

InChI key

YHHVXVNXTMIXOL-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luba Hunakova et al.
Toxicology letters, 279, 16-21 (2017-07-16)
Among a variety of metal containing organic compounds, tin derivatives are enjoying an increasing interest and appear to be very promising as potential drug candidates. We studied eight organometallic derivatives, nuclear retinoid X receptor (RXR) ligands and two germanium containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service