Merck
  • Sn- and Ge- triorganometallics exert different cytotoxicity and modulation of migration in triple-negative breast cancer cell line MDA-MB-231.
Sn- and Ge- triorganometallics exert different cytotoxicity and modulation of migration in triple-negative breast cancer cell line MDA-MB-231.
Toxicology letters (2017-07-16)
Luba Hunakova, Julius Brtko
ABSTRACT
Among a variety of metal containing organic compounds, tin derivatives are enjoying an increasing interest and appear to be very promising as potential drug candidates. We studied eight organometallic derivatives, nuclear retinoid X receptor (RXR) ligands and two germanium containing derivates that do not serve as RXR ligands. Tributylgermanium chloride (TBGe) and triphenylgermanium chloride (TPGe) did not inhibit growth of human triple negative MDA-MB-231 breast cancer cells. On the other hand, the tributyltin derivatives were highly, the triphenyltin derivatives less cytotoxic, both groups with IC
Sigma-Aldrich
Triphenylgermanium chloride, 99%
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder