Skip to Content
Merck
All Photos(1)

Key Documents

11318

Sigma-Aldrich

5-Amino-1-pentanol solution

50% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)5OH
CAS Number:
Molecular Weight:
103.16
Beilstein:
1732302
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

concentration

50% in H2O

bp

122 °C/16 mmHg (lit.)

density

0.99 g/mL at 20 °C

SMILES string

NCCCCCO

InChI

1S/C5H13NO/c6-4-2-1-3-5-7/h7H,1-6H2

InChI key

LQGKDMHENBFVRC-UHFFFAOYSA-N

General description

5-Aminopentan-1-ol is a colorless to faint yellow liquid.

Application

5-amino-1-pentanol is used in the preparation of fibres from random copolypeptides consisting of N-hydroxyalkyl L-glutamine and gamma-methyl L-glutamate with different monomer ratios. 5-amino-1-pentanol (5-Aminopentan-1-ol) was also used in the preparation of 5-phthalimidopentanal and tetracyclic core of the natural compound (+/-)-dibromoagelaspongin.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvain Picon et al.
Organic letters, 11(12), 2523-2526 (2009-05-19)
A six-step synthesis of the tetracyclic core of the natural compound (+/-)-dibromoagelaspongin, isolated from Agelas sp. Sponge, was achieved from the commercially available 5-aminopentan-1-ol, 2-trichloroacetylpyrrole, and 2-aminopyrimidine. Following a biomimetic inspired approach, successive oxidative reactions including the final DMDO biomimetic
T Hayashi et al.
Biomaterials, 11(6), 409-413 (1990-08-01)
Fibres from random copolypeptides consisting of N-hydroxyalkyl L-glutamine and gamma-methyl L-glutamate with different monomer ratios were prepared by aminoalcoholysis of a poly(gamma-methyl L-glutamate) fibre with 2-amino-1-ethanol or 5-amino-1-pentanol, both with 1,8-octamethylenediamine as cross-linking agent. The initial tensile properties as well
Miller D, et al.
Journal of the Chemical Society. Perkin Transactions 1, 1, 131-142 (1998)
Ahmed A Eltoukhy et al.
ACS nano, 8(8), 7905-7913 (2014-07-11)
Despite intensive research effort, the rational design of improved nanoparticulate drug carriers remains challenging, in part due to a limited understanding of the determinants of nanoparticle entry and transport in target cells. Recent studies have shown that Niemann-Pick C1 (NPC1)
Yohei Ishibashi et al.
Biochemical and biophysical research communications, 452(4), 873-880 (2014-08-27)
Galactosylglycerolipids (GGLs) and chlorophyll are characteristic components of chloroplast in photosynthetic organisms. Although chlorophyll is anchored to the thylakoid membrane by phytol (tetramethylhexadecenol), this isoprenoid alcohol has never been found as a constituent of GGLs. We here described a novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service