Merck
  • Biomimetically inspired short access to the 2-aminoimidazole-fused tetracyclic core of (+/-)-dibromoagelaspongin.
Biomimetically inspired short access to the 2-aminoimidazole-fused tetracyclic core of (+/-)-dibromoagelaspongin.
Organic letters (2009-05-19)
Sylvain Picon, Huu Dau Elise Tran, Marie-Thérèse Martin, Pascal Retailleau, Anne Zaparucha, Ali Al-Mourabit
ABSTRACT
A six-step synthesis of the tetracyclic core of the natural compound (+/-)-dibromoagelaspongin, isolated from Agelas sp. Sponge, was achieved from the commercially available 5-aminopentan-1-ol, 2-trichloroacetylpyrrole, and 2-aminopyrimidine. Following a biomimetic inspired approach, successive oxidative reactions including the final DMDO biomimetic oxidation gave the interesting triaminomethane-fused core.