Z363561
Indicator dots, “Radiation”
pkg of 1000 dots per roll
About This Item
Código UNSPSC:
26142300
NACRES:
NB.35
Productos recomendados
fabricante / nombre comercial
Bel-Art
envase
pkg of 1000 dots per roll
Descripción general
Adhesive dots, 1/4 in. diameter, with the universal "radiation" pictogram in red on yellow background. Color coding complies with OSHA regulations. Use on microcentrifuge tubes, gel sandwiches, anywhere a radiation warning is needed. There are 1,000 dots per roll.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)