Acerca de este artículo
NACRES:
NB.12
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific 120835
Feature:
to hold 120 x 15 mL tubes or 20 mm round bottom tubes
Material:
blue polypropylene
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarlematerial
blue polypropylene
feature
to hold 120 x 15 mL tubes or 20 mm round bottom tubes
manufacturer/tradename
Heathrow Scientific 120835
L × W × H
377 mm × 301 mm × 91 mm
General description
Fully assembled, polypropylene mega racks are great for high throughput labs requiring large sample storage. Each rack has 120 wells and a well diameter of 20 mm.
Features and Benefits
- Sturdy, durable material for easy and secure transport of clinical and research tubes
- Alphanumeric identification for quick location and retrieval of tubes
- Reusable, easy to clean and Autoclavable
- Place different test groupings into one rack eliminating the need for multiple racks
- Stackable for staging and saving space
Legal Information
Mega Rack is a registered trademark of Heathrow Scientific
Still not finding the right product?
Explore all of our products under Mega Rack® tube rack
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?