Documentos clave
EP5409709003
Lid for Eppendorf(R) F-45-30-11 Rotor
non aerosol tight, for 5427R, polypropylene
About This Item
UNSPSC Code:
41103913
NACRES:
NB.82
material
polypropylene
feature
non aerosol tight
manufacturer/tradename
Eppendorf® 5409709003 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with 5427 R
General description
Lids to fit the Eppendorf rotor F-45-30-11.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG