Skip to Content
Merck
All Photos(1)

Key Documents

PZ0318

Sigma-Aldrich

PF-06761281

≥97% (HPLC)

Synonym(s):

(2R)-2-Hydroxy-2-[2-(2-methoxy-5-methyl-3-pyridinyl)ethyl]-butanedioic acid, (R)-2-Hydroxy-2-(2-(2-methoxy-5-methylpyridin-3-yl)ethyl)-succinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO6
CAS Number:
Molecular Weight:
283.28
MDL number:
UNSPSC Code:
12161503
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1=CC(CC[C@](CC(O)=O)(O)C(O)=O)=C(OC)N=C1

InChI

1S/C13H17NO6/c1-8-5-9(11(20-2)14-7-8)3-4-13(19,12(17)18)6-10(15)16/h5,7,19H,3-4,6H2,1-2H3,(H,15,16)(H,17,18)/t13-/m1/s1

InChI key

FGYMJXFSHBLHLW-CYBMUJFWSA-N

General description

PF-06761281 is a small, polar dicarboxylate compound. Its structure is similar to citrate, which is the endogenous substrate of Na+-citrate cotransporter (NaCT). It is a novel, potent inhibitor of SLC13a2/3/5 (solute channel) family, along with selectivity for NaCT.

Application

PF-06761281 has been used as a SLC13A family inhibitor, in rats to determine its unbound partition coefficient (Kpuu), to estimate its potency as a drug.

Biochem/physiol Actions

PF-06761281 is an inhibitor of the sodium-coupled citrate transporter (NaCT or SLC13A5), which may be a target for tretment and prevention of metabolic disorders. The SLC13 transporters SLC13A2 (NaDC1), SLC13A3 (NaDC3), and SLC13A5 (NaCT) co-transport di- and tricarboxylates with multiple sodium ions into cells. PF-06761281 inhibits citrate uptake with an IC50 of 740 nM for NaCT in human hepatocytes. PF-06761281 has >25-fold in vitro selectivity for NaCT over NaDC1 and NaDC3 and was inactive in a selectivity panel of 65 targets.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Unbound Partition Coefficient and in Vitro-in Vivo Extrapolation for SLC13A Transporter-Mediated Uptake.
Riccardi K et al
Drug Metabolism and Disposition, 44(10), 1633-1642 (2016)
Kim Huard et al.
Journal of medicinal chemistry, 59(3), 1165-1175 (2016-01-07)
Inhibition of the sodium-coupled citrate transporter (NaCT or SLC13A5) has been proposed as a new therapeutic approach for prevention and treatment of metabolic diseases. In a previous report, we discovered dicarboxylate 1a (PF-06649298) which inhibits the transport of citrate in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service