Skip to Content
Merck
All Photos(1)

Key Documents

CLS8508

Corning® storage tube racks

empty storage rack w/ lids, for 96 screw capped or thermoplastic elastomers (TPE) capped storage tubes, 5 racks/pack, 2 packs/case, 10 racks/case

Synonym(s):

96 tube storage racks, empty storage racks, storage racks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.13
Pricing and availability is not currently available.

material

polypropylene

sterility

non-sterile

feature

Holds 96 x 1.3 mL tubes

packaging

case of 10
pack of 5

manufacturer/tradename

Corning 8508

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

  • These polypropylene racks are designed to hold 25 Corning 96 1.3mL storage tubes
  • For use with Cat. Nos. CLS6500 and CLS8502

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service