Z569860
2mag MIXcontrol 20 control unit for MIXdrive
AC/DC input 100 - 240 V AC, US/Japan plug
Synonym(s):
control unit, power bank, power unit
All Photos(1)
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Recommended Products
material
stainless steel plate
packaging
pack of 1 ea
manufacturer/tradename
2mag 90200
parameter
100-1600 rpm speed
Related Categories
Features and Benefits
- Control unit with 20 watts power
- Speed range from 100 up to 1,600 rpm
- Jerk-free stirring also at low speeds
- Power setting (4-steps) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation of MIXdrive stirring drives
- Clear digital display for speed- and power settings
- SoftStart for reliable catching/centering and safe accelerating of the stirring bar
- QuickSet for quick setting of start- and maximum speed, automatic memory function to save the last settings, resistant stainless steel housing, easy to clean
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)