A propos de cet article
NACRES:
NB.17
UNSPSC Code:
41103032
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aidermanufacturer/tradename
Diversified Biotech TNY-WTR
color
black
General description
A retractable Sharpie® pen with an extra fine tip. Recommended for writing on all our labels for samples that will be stored at room temperature and not exposed to chemicals and/or caustic agents.
Legal Information
Sharpie is a registered trademark of Sanford, LP
Still not finding the right product?
Explore all of our products under Teeny writer laboratory marker