Principaux documents
T1068
Pharmed L/S tubing
size 18
Synonyme(s) :
Saint Gobain AY242022, Peristaltic Pump Tubing
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.35
Conditionnement
pack of 25 feet
Fabricant/nom de marque
Tygon
D.I.
0.31 in. (7.9 mm)
Description générale
Tubing for MasterFlex Peristaltic Pumps, PharMed tubing has been specifically formulated for the MasterFlex pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure; reduces tubing cost and pump downtime.
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Heat sealable, bondable, and formable
  • Extremely low gas permeability
Informations légales
MasterFlex is a trademark of Cole-Parmer Instrument Co.