Principaux documents
MTCR4015
MTC Bio CoolCaddy Cold Station
to hold, PCR plate, tubes and cryovials
Synonyme(s) :
PCR cooler, benchtop cooler, sample tube
About This Item
Code UNSPSC :
41103037
Nomenclature NACRES :
NA.21
Matériaux
acrylonitrile-butadiene-styrene (ABS)
Caractéristiques
to hold PCR plate, tubes and cryovials
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio R4015
Description générale
CoolCaddy keeps your reagents and samples cool on the lab bench without using crushed ice. Simply store your CoolCaddy overnight in any standard freezer and it’s ready to hold temperatures between 2° and 4°C for up to 5 hours. Tubes, plates, strips and multi-channel reservoir solutions can all be kept cool while you perform your experiments.
Caractéristiques et avantages
  • Maintains 2-4°C for up to 5 hours
  • Keeps samples cool, but not frozen
  • Accommodates several different vessel types
  • 1.5/2.0mL Microtubes: 9 Tubes
  • 2.0mL Cryotubes: 8 Tubes
  • 0.5mL Microtubes: 5 Tubes
  • PCR Plates: 1 Plate
  • PCR Tubes: 96 Tubes or 12 Strips
  • Reservoirs: 1 x 50mL Reservoir
Informations légales
CoolCaddy is a trademark of MTC Bio
MTC is a trademark of MTC Bio