Principaux documents
HS8882A
Vinyl-Coated Lead Ring ("C" shape)
fits beaker size, 125 to 500 mL, blue
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
blue
Caractéristiques
fits beaker size 125 to 500 mL
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Heathrow Scientific HS8882A
Description générale
The Lead Ring is perfect for stabilizing vessels and labware or for water bath immersions.
Caractéristiques et avantages
  • Vinyl-coated lead ring will not mark or scratch benchtops and labware
  • O-Shape Style