Principaux documents
EP5910757000
Adapter for Eppendorf® S-4x Universal Large Rotor
for 750 mL conical tubes, pkg of 2 ea
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.82
Caractéristiques
Holds 750 mL flat bottom bottles
for 750 mL conical tubes
Conditionnement
pkg of 2 ea
Fabricant/nom de marque
Eppendorf® 5910757000
Technique(s)
centrifugation: suitable
Compatibilité
for use with Universal bucket
Description générale
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG