Principaux documents
EP5427757007
Rotor for Eppendorf® 5430/5430R Centrifuges
S-24-11-AT, aerosol-tight, to hold, 24 x 1.5/2.0 mL tubes, with lid and wind shield
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 24 x 1.5/2.0 mL tubes
aerosol-tight
swing bucket rotor
with lid and wind shield
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5427757007 (Global)
Paramètres
12,700 rpm
16,050 × g max. RCF
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models 5430/5430R
Description générale
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG