Key Documents
Z156515
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 28/12
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
Matériaux
stainless steel
Joint
joint: SJ28/12
Description générale
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.