Key Documents
Z118346
Cotton blend glove liners
size M/L, light weight
Synonyme(s) :
Cotton Glove Liners, resuable fabric glove
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
Matériaux
(100%Cotton)
white gloves
Taille
M/L
Stérilité
non-sterile
Caractéristiques
latex free
powder-free
reusable
Conditionnement
pack of 12 pair
Fabricant/nom de marque
Sigma-Aldrich
Longueur
9 in.
Description générale
Use these gloves for product protection during inspection or light manufacturing. Also ideal for use as a liner underneath outer gloves. Reversible pattern allows glove to be worn on either hand. Lightweight styles feature unhemmed cuffs.